Merck
  • Long Stress Induced Non-Coding Transcripts 5 (LSINCT5) Promotes Hepatocellular Carcinoma Progression Through Interaction with High-Mobility Group AT-hook 2 and MiR-4516.
Long Stress Induced Non-Coding Transcripts 5 (LSINCT5) Promotes Hepatocellular Carcinoma Progression Through Interaction with High-Mobility Group AT-hook 2 and MiR-4516.
Medical science monitor : international medical journal of experimental and clinical research (2018-11-26)
Ou Li, Zhizhen Li, Qinghe Tang, Yesheng Li, Shengxian Yuan, Yang Shen, Zhenhua Zhang, Nianfeng Li, Kaijian Chu, Guanglin Lei
ABSTRACT
BACKGROUND Long non-coding RNAs (lncRNAs) have been implicated in various human cancer types. However, the underlying mechanisms involved in hepatocellular carcinoma (HCC) progression remain poorly understood. MATERIAL AND METHODS In this study, lncRNA array was used to identify HCC related lncRNAs. RNA immunoprecipitation (RIP) followed mass spectrometry was used to explore lncRNA binding proteins. Western blot, quantitative PCR, tumor sphere formation, migration and viability assay were performed to evaluate the oncogenic role of lncRNAs. RESULTS We identified a novel lncRNA named long stress induced non-coding transcripts 5 (LSINCT5) which facilitates HCC progression. LSINCT5 was significantly upregulated in both HCC specimens and cell lines and correlates with poor survival. In vitro experiments showed that LSINCT5 promoted migration and viability of HepG2 and Huh7 cells. The in vivo xenograft mouse model also confirmed an oncogenic role for LSINCT5. RIP in combination with mass spectrometry identified HMGA2 as the LSINCT5 binding partner. LSINCT5 could bind to HMGA2 and decrease proteasome-mediated HMGA2 degradation leading to EMT activation. LSINCT5 also served as a competing endogenous RNA (ceRNA) for miR-4516, resulting in increased STAT3/BclxL expression and attenuated apoptosis. CONCLUSIONS Our data have collectively established a lncRNA LSINCT5 mediated process during HCC carcinogenesis and might have provided novel insight into therapeutic targeting.
Sigma-Aldrich
Monoclonal Anti-GAPDH antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL3266, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
(R)-MG132
Sigma-Aldrich
Anti-HMGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Ubiquitin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-GSK3B antibody produced in mouse, clone 3D10, ascites fluid
Sigma-Aldrich
Anti-STAT3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution