Merck
  • NADPH oxidase is the major source of placental superoxide in early pregnancy: association with MAPK pathway activation.
NADPH oxidase is the major source of placental superoxide in early pregnancy: association with MAPK pathway activation.
Scientific reports (2019-09-29)
Isabelle Hernandez, Thierry Fournier, Audrey Chissey, Patrice Therond, Abdel Slama, Jean-Louis Beaudeux, Amal Zerrad-Saadi
ABSTRACT
First-trimester placenta (<10 gestational weeks (GW)) develops in a low oxygen environment (≈2%). Early oxygen exposure can cause oxidative damage leading to pregnancy disorders. The aim of this work was to determine the major sources of placental superoxide during early pregnancy - more specifically before 10 GW - and to study redox adaptation to increased oxygen pressure after 12 GW. Our results show that NADPH oxidase (Nox) is the main source of superoxide in first-trimester chorionic villi. Its activity is higher before 10 GW and concomitant with the location on the syncytiotrophoblast apical pole of p47phox, the Nox organizer subunit. After the increase in pO2 pressure (12-14 GW), the activities of the antioxidant enzymes SOD1, catalase and GPX1 are increased. The redox-sensitive MAPK pathways show increased phosphorylated-p38 expression, but no variation in the phosphorylation of stress-activated protein kinase/c-Jun NH2-terminal kinase (SAPK/JNK) during first trimester, suggesting a physiological redox adaptation, whilst ERK1/2 phosphorylation is higher after 12 GW. Nox is the major superoxide source in early pregnancy (<10 GW). Increased superoxide production at 7-9 GW is associated with p38 MAPK pathway activation, suggesting that it is involved in physiological placental function and healthy early development of the placenta, through MAPK pathways.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
Anti-Keratin 7 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Phalloidin–Atto 550, suitable for fluorescence
Sigma-Aldrich
Anti-p47-phox Antibody, serum, Upstate®