Merck
  • Design, development, synthesis, and docking analysis of 2'-substituted triclosan analogs as inhibitors for Plasmodium falciparum enoyl-ACP reductase.
Design, development, synthesis, and docking analysis of 2'-substituted triclosan analogs as inhibitors for Plasmodium falciparum enoyl-ACP reductase.
IUBMB life (2009-10-28)
Neha Kapoor, Tanushree Banerjee, Ponnusamy Babu, Koustav Maity, Namita Surolia, Avadhesha Surolia
ABSTRACT
A structure-based approach has been adopted to develop 2'-substituted analogs of triclosan. The Cl at position 2' in ring B of triclosan was chemically substituted with other functional groups like NH(2), NO(2) and their inhibitory potencies against PfENR were determined. The binding energies of the 2' substituted analogs of triclosan for enoyl-acyl carrier protein reductase (ENR) of Plasmodium falciparum were determined using Autodock. Based on the autodock results, we synthesized the potential compounds. The IC(50) and inhibition constant (K(i)) of 2' substituted analogs of triclosan were determined against purified PfENR. Among them, two compounds, 2-(2'-Amino-4'-chloro-phenoxy)-5-chloro-phenol (compound 4) and 5-chloro-2-(4'-chloro-2'-nitro-phenoxy)-phenol) (compound 5) exhibited good potencies. Compound 4 followed uncompetitive inhibition kinetics with crotonoyl CoA and competitive with NADH. It was shown to have an IC(50) of 110 nM; inhibition constant was 104 nM with the substrate and 61 nM with the cofactor. IC(50) of compound 5 was determined to be 229 nM. Compounds 4 and 5 showed significant inhibition of the parasite growth in P. falciparum culture.
Sigma-Aldrich
2-Butenoyl coenzyme A lithium salt, ≥90% (HPLC)
Sigma-Aldrich
Crotonoyl coenzyme A trilithium salt, ~90% (HPLC)