Merck
  • Basal activity of a PARP1-NuA4 complex varies dramatically across cancer cell lines.
Basal activity of a PARP1-NuA4 complex varies dramatically across cancer cell lines.
Cell reports (2014-09-10)
Kristin A Krukenberg, Ruomu Jiang, Judith A Steen, Timothy J Mitchison
ABSTRACT
Poly(ADP-ribose) polymerases (PARPs) catalyze poly(ADP-ribose) addition onto proteins, an important posttranslational modification involved in transcription, DNA damage repair, and stem cell identity. Previous studies established the activation of PARP1 in response to DNA damage, but little is known about PARP1 regulation outside of DNA repair. We developed an assay for measuring PARP activity in cell lysates and found that the basal activity of PARP1 was highly variable across breast cancer cell lines, independent of DNA damage. Sucrose gradient fractionation demonstrated that PARP1 existed in at least three biochemically distinct states in both high- and low-activity lines. A discovered complex containing the NuA4 chromatin-remodeling complex and PARP1 was responsible for high basal PARP1 activity, and NuA4 subunits were required for this activity. These findings present a pathway for PARP1 activation and a direct link between PARP1 and chromatin remodeling outside of the DNA damage response.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-TIP60 Rabbit pAb, liquid, Calbiochem®