Merck
  • Fatty acid 2-hydroxylase regulates cAMP-induced cell cycle exit in D6P2T schwannoma cells.
Fatty acid 2-hydroxylase regulates cAMP-induced cell cycle exit in D6P2T schwannoma cells.
Journal of lipid research (2009-01-28)
Nathan L Alderson, Hiroko Hama
ABSTRACT
Sphingolipids are ubiquitous components of eukaryotic cells that regulate various cellular functions. In many cell types, a fraction of sphingolipids contain 2-hydroxy fatty acids, produced by fatty acid 2-hydroxylase (FA2H), as the N-acyl chain of ceramide [hydroxyl fatty acid (hFA)-sphingolipids]. FA2H is highly expressed in myelin-forming cells of the nervous system and in epidermal keratinocytes. While hFA-sphingolipids are thought to enhance the physical stability of specialized membranes produced by these cells, physiological significance of hFA-sphingolipids in many other cell types is unknown. In this study, we report novel roles for FA2H and hFA-sphingolipids in the regulation of the cell cycle. Treatment of D6P2T Schwannoma cells with dibutyryl-cAMP (db-cAMP) induced exit from the cell cycle with concomitant upregulation of FA2H. Partial silencing of FA2H in D6P2T cells resulted in 60-70% reduction of hFA-dihydroceramide and hFA-ceramide, with no effect on nonhydroxy dihydroceramide and ceramide. Under these conditions, db-cAMP no longer induced cell cycle exit, and cells continued to grow and divide. Immunoblot analyses revealed that FA2H silencing prevented db-cAMP-induced upregulation of cyclin-dependent kinase inhibitors p21 and p27. These results provide evidence that FA2H is a negative regulator of the cell cycle and facilitates db-cAMP-induced cell cycle exit in D6P2T cells.
Product Description
Avanti
18:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860829P, powder
Avanti
18:1(2R-OH) Ceramide, Avanti Research - A Croda Brand 860827P, powder
Avanti
22:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860821P, powder
Avanti
22:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860822P, powder
Avanti
24:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860823P, powder
Avanti
12:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860812P, powder
Avanti
16:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860816P, powder
Avanti
17:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860817P, powder
Avanti
17:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860818P, powder
Avanti
24:1(2R-OH) Ceramide, Avanti Research - A Croda Brand 860825P, powder
Avanti
20:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860820P, powder
Avanti
20:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860819P, powder
Avanti
18:1(2S-OH) Ceramide, Avanti Research - A Croda Brand 860828P, powder
Avanti
12:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860811P, powder
Avanti
18:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860830P, powder
Avanti
16:0(2R-OH) Ceramide, Avanti Research - A Croda Brand 860815P, powder
Avanti
24:0(2S-OH) Ceramide, Avanti Research - A Croda Brand 860824P, powder