Merck
The protein kinase SIK downregulates the polarity protein Par3.
Oncotarget (2018-02-22)
Michael Vanlandewijck, Mahsa Shahidi Dadras, Marta Lomnytska, Tanzila Mahzabin, Martin Lee Miller, Christer Busch, Søren Brunak, Carl-Henrik Heldin, Aristidis Moustakas
ABSTRACT
The multifunctional cytokine transforming growth factor β (TGFβ) controls homeostasis and disease during embryonic and adult life. TGFβ alters epithelial cell differentiation by inducing epithelial-mesenchymal transition (EMT), which involves downregulation of several cell-cell junctional constituents. Little is understood about the mechanism of tight junction disassembly by TGFβ. We found that one of the newly identified gene targets of TGFβ, encoding the serine/threonine kinase salt-inducible kinase 1 (SIK), controls tight junction dynamics. We provide bioinformatic and biochemical evidence that SIK can potentially phosphorylate the polarity complex protein Par3, an established regulator of tight junction assembly. SIK associates with Par3, and induces degradation of Par3 that can be prevented by proteasomal and lysosomal inhibition or by mutation of Ser885, a putative phosphorylation site on Par3. Functionally, this mechanism impacts on tight junction downregulation. Furthermore, SIK contributes to the loss of epithelial polarity and examination of advanced and invasive human cancers of diverse origin displayed high levels of SIK expression and a corresponding low expression of Par3 protein. High SIK mRNA expression also correlates with lower chance for survival in various carcinomas. In specific human breast cancer samples, aneuploidy of tumor cells best correlated with cytoplasmic SIK distribution, and SIK expression correlated with TGFβ/Smad signaling activity and low or undetectable expression of Par3. Our model suggests that SIK can act directly on the polarity protein Par3 to regulate tight junction assembly.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-GM130 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SIK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Partitioning-defective 3 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-PARD3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M5 antibody produced in mouse, clone M5, purified immunoglobulin, buffered aqueous solution