Skip to Content
Merck
Zirconium tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionate) ≥99.99%

Zirconium tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionate)

Zirconium tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionate) ≥99.99%
Synonym(s):
Zr(TMHD)4
Linear Formula:
Zr(OCC(CH3)3CHCOC(CH3)3)4
CAS Number:
Molecular Weight:
824.29

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing