
Ytterbium

Synonym(s):
Ytterbium
Empirical Formula (Hill Notation):
Yb
CAS Number:
Molecular Weight:
173.04
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (mm)
Element
Showing 1-15 of 15
Compare
Product Number
Description
Pricing