![Withanolide A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/239/765/fbd9a064-53e2-4324-8271-c2e6f372f03a/640/fbd9a064-53e2-4324-8271-c2e6f372f03a.png)
Withanolide A
![Withanolide A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/239/765/fbd9a064-53e2-4324-8271-c2e6f372f03a/640/fbd9a064-53e2-4324-8271-c2e6f372f03a.png)
Synonym(s):
(5α,6α,7α,22R)-6,7-Epoxy-5,20,22-trihydroxy-1-oxo-ergosta-2,24-dien-26-oic acid δ-lactone, Withaniol
Empirical Formula (Hill Notation):
C28H38O6
CAS Number:
Molecular Weight:
470.60
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3