
Vitamin B12a hydrochloride

Synonym(s):
Hydroxocobalamin hydrochloride
Empirical Formula (Hill Notation):
C62H89CoN13O15P · HCl
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing