
Vigabatrin Related Compound A

Synonym(s):
(5RS)-5-Ethenylpyrrolidin-2-one, 5-Vinyl-2-pyrrolidinone
Empirical Formula (Hill Notation):
C6H9NO
CAS Number:
Molecular Weight:
111.14
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2