
Ursodeoxycholic acid for system suitability

Synonym(s):
Ursodeoxycholic acid, 3α,7β-Dihydroxy-5β-cholan-24-oic acid, 5β-Cholan-24-oic acid-3α,7β-diol, 7β-Hydroxylithocholic acid, UDCS
Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing