
Triethylene glycol

Synonym(s):
Triethylene glycol, Triglycol
Linear Formula:
HO(CH2CH2O)2CH2CH2OH
CAS Number:
Molecular Weight:
150.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing