Skip to Content
Merck
Triclosan certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Triclosan

Triclosan certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Triclosan, 5-Chloro-2-(2,4-dichlorophenoxy)phenol, Irgasan
Empirical Formula (Hill Notation):
C12H7Cl3O2
CAS Number:
Molecular Weight:
289.54
EC Number:

Products

Product Category
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique