
trans-Caffeic acid

Synonym(s):
trans-3,4-Dihydroxycinnamic acid
Empirical Formula (Hill Notation):
C9H8O4
CAS Number:
Molecular Weight:
180.16
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2