
Titanium(IV) oxide

Synonym(s):
Titania, Titanium dioxide, Titanium(IV) oxide
Linear Formula:
TiO2
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Showing 1-9 of 9
Compare
Product Number
Description
Pricing