
Tetramethylrhodamine isothiocyanate mixed isomers

Synonym(s):
MRITC, TRITC, Tetramethylrhodamine B isothiocyanate
Empirical Formula (Hill Notation):
C25H21N3O3S
CAS Number:
Molecular Weight:
443.52
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing