Skip to Content
Merck
Tetramethylammonium hydrogensulfate hydrate 98%

Tetramethylammonium hydrogensulfate hydrate

Tetramethylammonium hydrogensulfate hydrate 98%
Linear Formula:
(CH3)4N(HSO4) · xH2O
CAS Number:
Molecular Weight:
171.22 (anhydrous basis)

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing