
Testosterone Related Compound I

Synonym(s):
17β-Hydroxyandrosta-4,6-dien-3-one, 4,6-Androstadiene-17β-ol-3-one
Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing