
Tenofovir

Synonym(s):
[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid monohydrate
Empirical Formula (Hill Notation):
C9H14N5O4P · H2O
CAS Number:
Molecular Weight:
305.23
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing