
Tapentadol-D3 hydrochloride solution

Synonym(s):
Tapentadol-D3 hydrochloride solution
Empirical Formula (Hill Notation):
C14D3H20NO · HCl
CAS Number:
Molecular Weight:
260.82
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Research Area
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing