
Sulfamonomethoxine

Synonym(s):
4-Amino-N-(6-methoxy-4-pyrimidinyl)benzenesulfonamide, 4-Methoxy-6-sulfanilamidopyrimidine, N1-(6-Methoxy-4-pyrimidinyl)sulfanilamide, SMM
Empirical Formula (Hill Notation):
C11H12N4O3S
CAS Number:
Molecular Weight:
280.30
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing