
Sulfaguanidine

Synonym(s):
4-Amino-N-(aminoiminomethyl)benzenesulfonamide, 4-Amino-N-guanylbenzenesulfonamide
Empirical Formula (Hill Notation):
C7H10N4O2S
CAS Number:
Molecular Weight:
214.24
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing