
Stanozolol impurity B

Synonym(s):
Oxymetholone, 17β-Hydroxy-2-(hydroxymethylene)-17-methyl-5α-androstan-3-one, Anasterone
Empirical Formula (Hill Notation):
C21H32O3
CAS Number:
Molecular Weight:
332.48
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing