
Sodium silicate solution

Synonym(s):
Sodium trisilicate solution, Water glass
Linear Formula:
Na2O(SiO2)x · xH2O
Products
Product Category
Brand
Physical Form
Chemical Composition
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing