Skip to Content
AZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Snap Cap Vials
Products
Product Category
vials (9)
Manufacturer
Binding Type
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
BAF175750003
Snap Cap Vials,
capacity 11 mL
Expand
View Pricing
BAF175750013
Snap Cap Vials,
capacity 48 mL
Expand
View Pricing
BAF175750005
Snap Cap Vials,
capacity 19 mL
Expand
View Pricing
BAF175750015
Snap Cap Vials,
capacity 55 mL
Expand
View Pricing
BAF175750010
Snap Cap Vials,
capacity 37 mL
Expand
View Pricing
BAF175750030
Snap Cap Vials,
capacity 111 mL
Expand
View Pricing
BAF175750007
Snap Cap Vials,
capacity 26 mL
Expand
View Pricing
BAF175750040
Snap Cap Vials,
capacity 148 mL
Expand
View Pricing
BAF175750020
Snap Cap Vials,
capacity 74 mL
Expand
View Pricing