
(S)-(+)-2-Phenylbutyric acid

Linear Formula:
C2H5CH(C6H5)CO2H
CAS Number:
Molecular Weight:
164.20
Products
Product Category
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing