Skip to Content
Merck
(S)-(−)-1-Boc-2-tert-butyl-3-methyl-4-imidazolidinone 99%

(S)-(−)-1-Boc-2-tert-butyl-3-methyl-4-imidazolidinone

(S)-(−)-1-Boc-2-tert-butyl-3-methyl-4-imidazolidinone 99%
Synonym(s):
(S)-(−)-1-(tert-Butoxycarbonyl)-2-tert-butyl-3-methyl-4-imidazolidinone
Empirical Formula (Hill Notation):
C13H24N2O3
CAS Number:
Molecular Weight:
256.34

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing