
Ropivacaine Related Compound A

Synonym(s):
2,6-Dimethylaniline hydrochloride, 2,6-Xylidine hydrochloride, 2,6-Xylidinium chloride
Empirical Formula (Hill Notation):
C8H11N · HCl
CAS Number:
Molecular Weight:
157.64
Products
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2