
(R)-(+)-α,α-Diphenyl-2-pyrrolidinemethanol

Synonym(s):
α,α-Diphenyl-D-prolinol, (R)-2-(Diphenylhydroxymethyl)pyrrolidine
Empirical Formula (Hill Notation):
C17H19NO
CAS Number:
Molecular Weight:
253.34
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing