
Quercetin 3-O-α-L-arabinopyranoside

Synonym(s):
3′,4′,5,7-Tetrahydroxyflavone 3-O-α-L-arabinoside, 3-(α-L-Arabinopyranosyloxy)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-1-benzopyran-4-one, Feniculin, Foeniculin, Guaijaverin, Quercetin 3-O-α-L-arabinoside
Empirical Formula (Hill Notation):
C20H18O11
CAS Number:
Molecular Weight:
434.35
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing