
Prothioconazole-desthio

Synonym(s):
α-(1-Chlorocyclopropyl)-α-[(2-chlorophenyl)methyl]-1H-1,2,4-triazole-1-ethanol
Empirical Formula (Hill Notation):
C14H15Cl2N3O
CAS Number:
Molecular Weight:
312.19
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2