
Propylthiouracil

Synonym(s):
6-Propyl-2-thiouracil, 2,3-Dihydro-6-propyl-2-thioxo-4(1H)-pyrimidinone, 4-Hydroxy-2-mercapto-6-propylpyrimidine
Empirical Formula (Hill Notation):
C7H10N2OS
CAS Number:
Molecular Weight:
170.23
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade