
Probenecid

Synonym(s):
p-(Dipropylsulfamoyl)benzoic acid, Probenecid
Empirical Formula (Hill Notation):
C13H19NO4S
CAS Number:
Molecular Weight:
285.36
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-4 of 4