
Prilocaine

Synonym(s):
Prilocaine, (±)-2′-Methyl-2-(propylamino)propionanilide, N-(2-Methylphenyl)-2-(propylamino)propanamide
Empirical Formula (Hill Notation):
C13H20N2O
CAS Number:
Molecular Weight:
220.31
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3