
Phospholipase C, Phosphatidylinositol-specific from Bacillus cereus

Synonym(s):
1-Phosphatidyl-D-myo-inositol inositol phosphohydrolase, cyclic-phosphate forming, PI-PLC
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing