
Penicillin G procaine

Synonym(s):
Penicillin G procaine salt monohydrate
Empirical Formula (Hill Notation):
C16H18N2O4S1 · C13H20N2O2 · H2O
CAS Number:
Molecular Weight:
588.72
Products
Product Category
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing