
Pellicon® 3 Cassette with Biomax® Membrane

Products
Product Category
Brand
pH Value
Product Line
Length (cm)
Length (in)
Surface Area cm²
Quality Segment
Housing Material
Membrane Material
MWCO (kDa)
Technique
Showing 1-16 of 16
Compare
Product Number
Description
Pricing