
Palmitoyl coenzyme A lithium salt

Synonym(s):
n-Hexadecanoyl Coenzyme A, Hexadecanoyl coenzyme A
Empirical Formula (Hill Notation):
C37H65N7O17P3S · xLi+
CAS Number:
Molecular Weight:
1004.94 (free acid basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing