
Paclitaxel

Synonym(s):
Paclitaxel, Taxus sp., InSolution Paclitaxel, Taxus sp., Paclitaxel, Semi-Synthetic, Paclitaxel
Empirical Formula (Hill Notation):
C47H51NO14
CAS Number:
Molecular Weight:
853.91
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Agency
Format
Research Area
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing