
Oxycodone HCl

Synonym(s):
Oxycodone hydrochloride
Empirical Formula (Hill Notation):
C18H21NO4 · HCl
CAS Number:
Molecular Weight:
351.82
Products
Brand
Formula Weight
Agency
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing