
n-Tridecane

Synonym(s):
Tridecane, n-Tridecane
Linear Formula:
CH3(CH2)11CH3
CAS Number:
Molecular Weight:
184.36
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing