
N,N-Diethyl-p-phenylenediamine

Synonym(s):
4-Amino-N,N-diethylaniline
Linear Formula:
(C2H5)2NC6H4NH2
CAS Number:
Molecular Weight:
164.25
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing