
N-Methyl-N′-(2,4-xylyl)formamidine

Synonym(s):
Amitraz Metabolite BTS 27271, N-(2,4-Dimethylphenyl)-N′-methylformamidine, N-Methyl-N′-2,4-xylylformamidine
Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing