Skip to Content
Merck
Nitrilotriacetic acid trisodium salt monohydrate ≥98.0% (KT)

Nitrilotriacetic acid trisodium salt monohydrate

Nitrilotriacetic acid trisodium salt monohydrate ≥98.0% (KT)
Synonym(s):
Sodium nitrilotriacetate tribasic monohydrate
Linear Formula:
N(CH2COONa)3 · H2O
CAS Number:
Molecular Weight:
275.10
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing