
Nicotine Related Compound E

Synonym(s):
(2S)-3-(1-Methyl-2-pyrrolidinyl)pyridine N-oxide oxalate salt, (1RS,2S)-1-Methyl-2-(pyridin-3-yl)pyrrolidine 1-oxide oxalate, Nicotine 1′-N-oxide oxalate salt, Oxynicotine oxalate
Empirical Formula (Hill Notation):
C10H14N2O · C2H2O4
CAS Number:
Molecular Weight:
268.27
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2