Skip to Content
Merck
Nickel(II) bromide hydrate 98%

Nickel(II) bromide hydrate

Nickel(II) bromide hydrate 98%
Linear Formula:
NiBr2 · xH2O
CAS Number:
Molecular Weight:
218.50 (anhydrous basis)

Products

Formula Weight
Physical Form
Purity
Chemical Composition
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing