
N-(2-Pyridyl)bis(trifluoromethanesulfonimide)

Synonym(s):
2-[N,N-Bis(trifluoromethylsulfonyl)amino]pyridine, N,N-Bis(trifluoromethylsulfonyl)-2-pyridylamine
Empirical Formula (Hill Notation):
C7H4F6N2O4S2
CAS Number:
Molecular Weight:
358.24
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing