
N-(2-Hydroxyethyl)aniline

Synonym(s):
2-(Phenylamino)ethanol, 2-Anilinoethanol, N-Phenylethanolamine
Linear Formula:
C6H5NHCH2CH2OH
CAS Number:
Molecular Weight:
137.18
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing