
N-(2-Chloroethyl)-N-ethyl-2-bromobenzylamine hydrochloride

Synonym(s):
DSP-4, N-Ethyl-N-(2-chloroethyl)-2-bromobenzylamine hydrochloride
Empirical Formula (Hill Notation):
C11H15BrClN · HCl
CAS Number:
Molecular Weight:
313.06
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing